Learn More
Alkali Scientific SureSeal™ 2.0mL Screw Cap Microtube with Black Silicone O-ring, Caps iIncluded, Self-standing, Sterile, Graduated, RNase, Dnase, Pyrogen free, Assembled, 50 per resealable bag, 1000/cs
SDP
Catalog No. 502336025
Encompass_Preferred
ClearSeal screw-cap microtubes come sterile in 2.0mL, ultra-clear polypropylene, screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal screw cap microtubes are sterile, RNase, DNase, and pyrogen-free and may be centrifuged at up to 22,000xg*. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.
Catalog No. 50-233-6025 Supplier Alkali Scientific Supplier No. MT3220SG
ClearSeal screw-cap microtubes come sterile in 2.0mL, ultra-clear polypropylene, screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal screw cap microtubes are sterile, RNase, DNase, and pyrogen-free and may be centrifuged at up to 22,000xg*. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.