Learn More
Alkali Scientific SureSeal™ 2.0mL with Loop Caps, Sterile, Graduated, with O-ring, Self-standing, Caps, Assembled, 50 per Resealable Bag
SDP
Catalog No. 502389047
Encompass_Preferred
ClearSeal screw-cap microtubes come sterile in 2.0mL, ultra-clear polypropylene, screw-cap microtubes with loop attached cap come with assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal screw cap microtubes are sterile, RNase, DNase, and pyrogen-free and may be centrifuged at up to 22,000xg*. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.
Catalog No. 50-238-9047 Supplier Alkali Scientific Supplier No. MT3220SGL
ClearSeal screw-cap microtubes come sterile in 2.0mL, ultra-clear polypropylene, screw-cap microtubes with loop attached cap come with assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal screw cap microtubes are sterile, RNase, DNase, and pyrogen-free and may be centrifuged at up to 22,000xg*. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.