Learn More
Molecular Dimensions SURESEAL DWB MD6-16
SDP
Catalog No. NC0382853
Encompass
Protein Crystallography. SureSeal Deep Well Block (100) Provides longer lasting seal for crystallization screen storage in deep-well blocks. Two end tabs. No residue left behind when removed. 137mm x 80mm sheet with 117mm x 80mm adhesive area.
Catalog No. NC0382853 Supplier Molecular Dimensions Supplier No. MD616
Protein Crystallography. SureSeal Deep Well Block (100) Provides longer lasting seal for crystallization screen storage in deep-well blocks. Two end tabs. No residue left behind when removed. 137mm x 80mm sheet with 117mm x 80mm adhesive area.