Learn More
Alkali Scientific SureSeal™ Transport Tubes, with O Ring Sealing Cap, 1000/Cs, 5mL, Separately Packaged O Ring
SDP
Catalog No. 5000662354
Encompass_Preferred
Transport tubes are used in nearly every type of laboratory, from medical to environmental to bio research. Often used to transport highly infectious samples, scientist have demanded the highest level of leak resistant containment. To meet that demand and other stress factors, an integral silicone O ring system has been developed to ensure safe containment in high/low temperature environments and during air transportation. Guaranteed to exceed 95kPa. Tubes come packaged in 10 bags of 100 tubes, with pre sterilized attached cap options, or non sterile with separately packaged caps. • Leak proof O ring seal • Exceeds 95kPa compliance standards • Graduations and marking spot • Sterile with attached caps • Non Sterile with separately packaged caps
Catalog No. 50-006-62354 Supplier Alkali Scientific Supplier No. C2813
Transport tubes are used in nearly every type of laboratory, from medical to environmental to bio research. Often used to transport highly infectious samples, scientist have demanded the highest level of leak resistant containment. To meet that demand and other stress factors, an integral silicone O ring system has been developed to ensure safe containment in high/low temperature environments and during air transportation. Guaranteed to exceed 95kPa. Tubes come packaged in 10 bags of 100 tubes, with pre sterilized attached cap options, or non sterile with separately packaged caps. • Leak proof O ring seal • Exceeds 95kPa compliance standards • Graduations and marking spot • Sterile with attached caps • Non Sterile with separately packaged caps