Learn More
Epredia™ Suresorb™ Disposable Dissecting Table Pads
Catalog No. 21124387 Shop All Epredia Products
Change view
Click to view available options
Description:
22 x 20 in.
Catalog No. Description
21-124-387 22 x 20 in.
1 options
Catalog No. 21-124-387 Supplier Epredia™ Supplier No. 9834
Add to Cart
Add to Cart
Absorb and lock in liquids with Epredia™ Suresorb™ Disposable Dissecting Table Pads.
  • Epredia offers a full range of organized, stand-alone, and benchtop pathology workstations for the gross examination band sectioning of specimens.
  • Clinical pads work as surgical cutting board pads, dissecting pads, autopsy table pads and liners for body trays and cadaver bags.
  • Fiber resists tearing, even when saturated
  • Ideal for grossing areas and autopsy work
Specifications
Description 22 x 20 in.
Type Absorbent Sheet