Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ SureStack Pipet Tips

Catalog No. 02100503 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Natural
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Color
02-100-503 200 μL Sterile Natural
02-100-500 10 μL Non-sterile Natural
02-100-505 200 μL Sterile Yellow
02-100-501 10 μL Sterile Natural
02-100-502 200 μL Non-sterile Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-100-503
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pipetting station with high quality tips.

  • Ideal for use with multi-channel pipetters, both 8 and 12 channel
  • Decks snap together and will not lift during pipetting
  • Recessed wells keep tips stable for mounting
  • Saves bench space
  • Lid can be adjusted to expose only 96 tips
  • Wide base design prevents stack from tipping
  • Trays manufactured from recycled plastic
  • Autoclaving not recommended: available presterilized
  • 10μL Long Reach tip features extended length (45mm) and graduation marks at 2, 5 and 10μL
  • 200μL pipet tip features a soft fit collar, graduation marks at 10, 50 and 100μL

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line SureStack
Material Recycled Plastic
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.