Learn More
Research Products International Corp SureStand Pipette Rack, Cap 5, 2 multi
SDP
Catalog No. 501977968
Encompass
Universal SureStand Pipette Rack accommodates all single and multi-channel pipettes. Racks feature patent pending "window slots" that conveniently fit and hold multi-channel pipettes securely in place. Durable rack is manufactured of sturdy 5mm acrylic and non-skid rubber feet allow additional stability.
  • Holds all popular pipettes
  • Ideal for multi-channel pipettes
  • Versatile design
Catalog No. 50-197-7968 Supplier Research Products International Corp Supplier No. 401111
Universal SureStand Pipette Rack accommodates all single and multi-channel pipettes. Racks feature patent pending "window slots" that conveniently fit and hold multi-channel pipettes securely in place. Durable rack is manufactured of sturdy 5mm acrylic and non-skid rubber feet allow additional stability.
  • Holds all popular pipettes
  • Ideal for multi-channel pipettes
  • Versatile design