Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureSTART™ 9 mm Screw Caps, Level 1 Everyday Analysis
GSA_VAGREENER_CHOICE

Catalog No. 03251835
Change view
Click to view available options
Septum:
Red Natural Rubber/Clear TEF
Red Silicone/Ivory PTFE
White Silicone/Blue PTFE
White Silicone/Red PTFE
White Silicone/White PTFE
Product Type:
AVCS Screw Cap
Screw Cap
Quantity:
100 Pk.
1000 Pk.
5000 Pk.
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum Product Type Quantity
03-251-835 Red Silicone/Ivory PTFE AVCS Screw Cap 100 Pk.
03-251-834 Red Natural Rubber/Clear TEF AVCS Screw Cap 100 Pk.
03-452-286 Red Natural Rubber/Clear TEF AVCS Screw Cap 1000 Pk.
03-452-292 Red Natural Rubber/Clear TEF AVCS Screw Cap 5000 Pk.
03-452-285 Red Silicone/Ivory PTFE AVCS Screw Cap 1000 Pk.
03-452-291 Red Silicone/Ivory PTFE AVCS Screw Cap 5000 Pk.
03-251-836 White Silicone/Red PTFE AVCS Screw Cap 100 Pk.
03-452-284 White Silicone/Red PTFE AVCS Screw Cap 1000 Pk.
03-452-290 White Silicone/Red PTFE AVCS Screw Cap 5000 Pk.
03-251-838 White Silicone/White PTFE Screw Cap 100 Pk.
03-452-322 White Silicone/White PTFE Screw Cap 1000 Pk.
03-452-328 White Silicone/White PTFE Screw Cap 5000 Pk.
03-251-839 White Silicone/White PTFE Screw Cap 100 Pk.
03-452-323 White Silicone/White PTFE Screw Cap 1000 Pk.
03-452-329 White Silicone/White PTFE Screw Cap 5000 Pk.
03-251-837 White Silicone/Blue PTFE AVCS Screw Cap 100 Pk.
03-452-287 White Silicone/Blue PTFE AVCS Screw Cap 1000 Pk.
03-452-293 White Silicone/Blue PTFE AVCS Screw Cap 5000 Pk.
01-549-804 White Silicone/Red PTFE Screw Cap 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 03-251-835 Supplier Thermo Scientific™ Supplier No. CHSC920
Only null left
Add to Cart
Edge
Add to Cart

Select Level 1 Thermo Scientific™ SureSTART™ 9 mm Screw Caps for chromatography approved, cost optimized products that are suitable for everyday analyses and compatible with all instrument types.

Use Thermo Scientific™ SureSTART™ 9 mm Screw Caps with screw vials that have a 9 mm opening, including our SureSTART 2 mL Glass Screw Vials and 2 mL Polypropylene Screw Microvials for <2 mL samples (Level 1). Select Level 1 caps for chromatography approved, cost optimized products that are suitable for everyday analyses and compatible with all instrument types. Choose from polypropylene caps with silicone/PTFE or rubber/TEF septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Our AVCS™ caps incorporate Advanced Vial Closure System technology to ensure optimal seal compression when closing a vial. Use AVCS screw caps with our SureStop™ vials for effective seals, consistent data, and increased sample throughput.

Our Performance Level 1 caps are cost-efficient and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses.

SureStop Vials and Advanced Vial Closure System (AVCS) Caps

This patented screw top vial/cap combination features a push-through resistant sealing ring design, controlled septum compression, and SureStop closure positioning flange to ensure an effective seal that eliminates under- and over-sealing, reduces solvent evaporation, and creates less autosampler interruptions. SureStop vials and AVCS caps have universal instrument compatibility for all LC and GC autosamplers. They also contain low leachable levels, making them an ideal alternative to bonded caps for those seeking cleaner spectra.

The SureSTART Performance Levels

SureSTART vials, caps, inserts, septa, and kits are available in three performance levels. Now you can choose the products that best suit your analytical, performance, or budget needs.

Level 1: Everyday analysis

Chromatography approved

SureSTART Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.

Level 2: High-throughput analysis

SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.

Level 3: High performance

When sensitivity is a must

SureSTART Level 3 products are packed in a clean environment according to our own hygiene concept and regulations to protect against contaminating factors. The vials contain the lowest levels of extractable metals and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes. The septa are conditioned to provide the lowest level of chromatographic extractables. Choose these products for ultimate method performance and high levels of and sample security and integrity.

All SureSTART vials, closures, and septa are certified to meet physical parameter specifications as part of our manufacturing process.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
Pre-slit No
Septum Red Silicone/Ivory PTFE
Level Level 1
Product Line SureSTART Level 1
Quantity 100 Pk.
Thickness (Metric) 1 mm
Closure Size 9 mm
For Use With (Equipment) 9 mm Screw Vials
Green Features Yes - Recyclable packaging
ACT Label Yes
Certifications/Compliance Specification Certified
Product Type AVCS Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.