Learn More
AUTOMATIONDIRECT COM INC SURESTEP DC ADVNCD MCRSTPPNG
Catalog No. NC3066521
Encompass
SureStep DC advanced microstepping stepper drive, 5A per phase,2-phase output, 24-48 VDC, bipolar, velocity and position mode,200 to 51200 steps per revolution, (1) RS-232 (RJ11) port(s).
Catalog No. NC3066521 Supplier AUTOMATIONDIRECT COM INC Supplier No. STPDRV4850
SureStep DC advanced microstepping stepper drive, 5A per phase,2-phase output, 24-48 VDC, bipolar, velocity and position mode,200 to 51200 steps per revolution, (1) RS-232 (RJ11) port(s).