Learn More
AUTOMATIONDIRECT COM INC SURESTEP STEPPER MOTOR
Catalog No. NC3055095
Encompass
SureStep stepper motor, NEMA 14 frame, IP40, single shaft, 0.35A,8 oz-in holding torque, 1.8-degree step angle, 200 steps perrevolution, bipolar. Requires STP-EXTL-xxx cable.
Catalog No. NC3055095 Supplier AUTOMATIONDIRECT COM INC Supplier No. STPMTRL14026
SureStep stepper motor, NEMA 14 frame, IP40, single shaft, 0.35A,8 oz-in holding torque, 1.8-degree step angle, 200 steps perrevolution, bipolar. Requires STP-EXTL-xxx cable.