Learn More
AUTOMATIONDIRECT COM INC SURESTEP STEPPER MOTOR NEMA17
Catalog No. NC3055100
Encompass
SureStep stepper motor, NEMA 17 frame, IP40, single shaft, 1.7A,61 oz-in holding torque, 1.8-degree step angle, 200 steps perrevolution, bipolar. Requires STP-EXT-xxx cable.
Catalog No. NC3055100 Supplier AUTOMATIONDIRECT COM INC Supplier No. STPMTR17040
SureStep stepper motor, NEMA 17 frame, IP40, single shaft, 1.7A,61 oz-in holding torque, 1.8-degree step angle, 200 steps perrevolution, bipolar. Requires STP-EXT-xxx cable.