Learn More
Alkali Scientific SureStand™™ Pipette Rack for Six Pipettes, and up to Four Multi-channels, 1/cs
SDP
Catalog No. 502335923
Encompass_Preferred
SureStand™ Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patent pending “windows” allow the user to insert the multi-channel head longways through the rack, while keeping the body of the pipette stable by securing the pipette at two points; under the finger hook, and near the bottom of the handle. SureStand™ is available in a 6-place rack that accepts four multi-channels. Each SureStand™ is also outfitted with four non-skid rubber “feet” for extra stability. SureStand™ is guaranteed to be the sturdiest and most versatile pipette rack in your lab.
Catalog No. 50-233-5923 Supplier Alkali Scientific Supplier No. P8406
SureStand™ Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patent pending “windows” allow the user to insert the multi-channel head longways through the rack, while keeping the body of the pipette stable by securing the pipette at two points; under the finger hook, and near the bottom of the handle. SureStand™ is available in a 6-place rack that accepts four multi-channels. Each SureStand™ is also outfitted with four non-skid rubber “feet” for extra stability. SureStand™ is guaranteed to be the sturdiest and most versatile pipette rack in your lab.