Learn More
Research Products International Corp SureStrain Premium Cell Strainers, 70µm Mesh, White, 50 per Case
SDP
Catalog No. 501977856
Encompass
SureStrain premium cell strainers are designed to offer faster isolation of cells from tissue or media. Advanced design features increased flow rate and improved aeration through nylon mesh. Molded grip ensures aseptic handling and eliminates contamination. Stackable strainers allow tiered filtration of different mesh sizes at one time and can also strain directly in petri dishes or plates.
Simply open the sterile package, attach the strainer to the top of a 50mL tube and add your cell suspension/sample. Or, attach the provided reducing adapter for use in 15ml tubes.
Catalog No. 50-197-7856 Supplier Research Products International Corp Supplier No. 162507
SureStrain premium cell strainers are designed to offer faster isolation of cells from tissue or media. Advanced design features increased flow rate and improved aeration through nylon mesh. Molded grip ensures aseptic handling and eliminates contamination. Stackable strainers allow tiered filtration of different mesh sizes at one time and can also strain directly in petri dishes or plates.
Simply open the sterile package, attach the strainer to the top of a 50mL tube and add your cell suspension/sample. Or, attach the provided reducing adapter for use in 15ml tubes.