Learn More
Hillrom SureTemp™ Plus 692 Thermometer
Catalog No. 22355164
Change view
Click to view available options
Battery Type:
3 Alkaline AA Batteries
Catalog No. Battery Type
22-355-164 3 Alkaline AA Batteries
1 options
This item is not returnable. View return policy
Catalog No. 22-355-164 Supplier Hillrom Supplier No. WEL 01692200
Add to Cart
Add to Cart
This item is not returnable. View return policy
SureTemp digital thermometer can be used as an oral thermometer, rectal thermometer and axillary thermometer. It has long been the gold standard in hand-held and integrated thermometer parameters, providing accurate, reliable readings.
  • Large LCD displays in Fahrenheit or Celsius
  • Convenient storage for 25 probe covers
  • Last temperature recall button
  • Security wall mount protects against theft
  • Captures about 6000 readings on 3 AA batteries
Specifications
Includes Interchangeable Oral Probe Well, Oral Probe
Length (English) Cable 4.0 ft.
Measurement Time 4 to 6 sec. Oral, 10 to 13 sec. Pediatric Axillary, 12 to 15 sec. Adult Axillary
Probe Type Oral
Product Type Thermometer
Battery Type 3 Alkaline AA Batteries
Display LCD