Learn More
Omega Engineering, Inc. SURF THERMOCPL W/ SLF ADHV 5PK
Catalog No. NC2588545
Encompass
Surface Thermocouple with Self-Adhesive Backing - Package of 5; Wire: 30 AWG PFA-coated; 80in length
Catalog No. NC2588545 Supplier Omega Engineering, Inc. Supplier No. SA1T72SC
Surface Thermocouple with Self-Adhesive Backing - Package of 5; Wire: 30 AWG PFA-coated; 80in length