Learn More
Aviva Systems Biology Corp SURF1 Antibody - N-terminal region (ARP61537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SURF1. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human
Catalog No. 50-287-403