Learn More
Aviva Systems Biology Corp SURF4 Antibody - N-terminal region (ARP50161_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50161_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SURF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SURF4. Alternate names: ERV29, FLJ22993, MGC102753ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-60273