Learn More
Aviva Systems Biology Corp SURF6 Antibody - middle region (ARP40681_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40681_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SURF6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SURF6. Alternate names: RRP14ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-57586