Learn More
Aviva Systems Biology Corp SURF6 Antibody - N-terminal region (ARP40680_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40680_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SURF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SURF6. Alternate names: RRP14ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-57585