Learn More
Apexbio Technology LLC surface antigen (208-215) [Hepatitis B Virus] 1mg
SDP
Catalog No. 5000095530
Encompass_Preferred
Surface antigen 208-215 Hepatitis B Virus is a peptide representing a defined epitope of the hepatitis B virus surface antigen (HBsAg) which is associated with host immune recognition and antibody-mediated responses The peptide encompasses antigenic determinants recognized during HBV infection facilitating studies focused on viral pathogenesis and immune interactions Surface antigen 208-215 Hepatitis B Virus exerts its biological activity through its defined antigenic sequence serving as a target for antibody detection and immunological assays Based on these properties surface antigen 208-215 Hepatitis B Virus holds research potential in virological studies immunological assay development diagnostics and vaccine research for hepatitis B virus
Catalog No. 50-000-95530 Supplier Apexbio Technology LLC Supplier No. A10911
Surface antigen 208-215 Hepatitis B Virus is a peptide representing a defined epitope of the hepatitis B virus surface antigen (HBsAg) which is associated with host immune recognition and antibody-mediated responses The peptide encompasses antigenic determinants recognized during HBV infection facilitating studies focused on viral pathogenesis and immune interactions Surface antigen 208-215 Hepatitis B Virus exerts its biological activity through its defined antigenic sequence serving as a target for antibody detection and immunological assays Based on these properties surface antigen 208-215 Hepatitis B Virus holds research potential in virological studies immunological assay development diagnostics and vaccine research for hepatitis B virus