Learn More
Grainger Surface Box White
Catalog No. 502766256
Encompass_Preferred
The Edwards Signaling 16X258 is a surface mount box for fire alarm systems. It is made of steel with a red powder coated finish and measures 4.5 x 4.5 x 2.75 inches. The box has a knock-out on one side and features mounting holes for easy installation. It can house the wiring connections for fire alarm devices and is UL listed. With its durable construction, it provides an enclosure to protect fire alarm wiring and components.
Catalog No. 50-276-6256 Supplier Grainger Supplier No. 16X258
The Edwards Signaling 16X258 is a surface mount box for fire alarm systems. It is made of steel with a red powder coated finish and measures 4.5 x 4.5 x 2.75 inches. The box has a knock-out on one side and features mounting holes for easy installation. It can house the wiring connections for fire alarm devices and is UL listed. With its durable construction, it provides an enclosure to protect fire alarm wiring and components.