Learn More
Grainger Surface Box White
Catalog No. 502765710
Encompass_Preferred
The Edwards Signaling Box is a white metal box with a hinged door and latch. It measures 16 inches wide by 12 inches high by 6 inches deep. The box is made of steel with a white powder coat finish. It has mounting holes to allow flush wall mounting. This box provides an enclosed space to house and protect electrical and electronic components. With its durable construction, it is well-suited for industrial, commercial, and residential applications requiring a secure metal enclosure. The latching door allows convenient access to the interior. Overall, this is a sturdy metal box for protecting electrical and electronic equipment in a range of settings.
Catalog No. 50-276-5710 Supplier Grainger Supplier No. 16X370
The Edwards Signaling Box is a white metal box with a hinged door and latch. It measures 16 inches wide by 12 inches high by 6 inches deep. The box is made of steel with a white powder coat finish. It has mounting holes to allow flush wall mounting. This box provides an enclosed space to house and protect electrical and electronic components. With its durable construction, it is well-suited for industrial, commercial, and residential applications requiring a secure metal enclosure. The latching door allows convenient access to the interior. Overall, this is a sturdy metal box for protecting electrical and electronic equipment in a range of settings.