Learn More
MSC ZEP Steam Cleaner, Heavy-Duty, Liquid, 55 gal Drum, 1 ea
Catalog No. 502817587
Encompass
Steam 'N Clean is a heavy-duty cleaner designed for use in high-pressure and steam cleaning equipment. It effectively removes paint, dirt, grease, and baked-on soil. It also strips many types of industrial and machinery paints and retards flash-rusting of cleaned surfaces.
  • Heavy-duty cleaner for high-pressure and steam cleaning
  • Rapidly removes paint, dirt, grease, and baked-on soil
  • Strips industrial and machinery paints
  • Retards flash-rusting on cleaned surfaces
  • Minimizes scale formation in steam-generation coils
  • Free-rinsing and essentially odorless
  • USDA applicable, A1
  • Performance unaffected by hard water
  • Does not contribute to heating coil scale
  • For iron, steel, stainless steel, and titanium parts
Catalog No. 50-281-7587 Supplier MSC Supplier No. 17733064
Steam 'N Clean is a heavy-duty cleaner designed for use in high-pressure and steam cleaning equipment. It effectively removes paint, dirt, grease, and baked-on soil. It also strips many types of industrial and machinery paints and retards flash-rusting of cleaned surfaces.
  • Heavy-duty cleaner for high-pressure and steam cleaning
  • Rapidly removes paint, dirt, grease, and baked-on soil
  • Strips industrial and machinery paints
  • Retards flash-rusting on cleaned surfaces
  • Minimizes scale formation in steam-generation coils
  • Free-rinsing and essentially odorless
  • USDA applicable, A1
  • Performance unaffected by hard water
  • Does not contribute to heating coil scale
  • For iron, steel, stainless steel, and titanium parts