Learn More
Texwipe Surface and Cleaning Validation Kit
SDP
Catalog No. 17377852 Shop All Texwipe Products
Change view
Click to view available options
Tip Style:
Broad
Catalog No. Tip Style
17-377-852 Broad
1 options
Catalog No. 17-377-852 Supplier Texwipe Supplier No. TX3360
Add to Cart
Add to Cart
Streamlines surface and cleaning validation by organizing swabs, vials, and labels in one secure kit
  • Organized, convenient and secure method for transporting swabs and vials from the storage area to sampling sites and laboratories with minimal chance of contamination
  • Doubled bagged to reduce the risk of contamination
  • Pre-washed borosilicate glass vials have caps with a one-piece cap-and-septum design for easy extraction of the vial content
  • Polyethylene over-cap protects septum surface from contamination until time of analysis
  • Includes TX718 Alpha™ Polyester Knit Large Sampling Swab
  • Cleanroom compatible for labels and traceability
  • Cleanroom compatible polypropylene box
Specifications
Product Type Test Swab
Sterility Non-sterile
Handle Material Polypropylene
Tip Style Broad
Tip Material Polyester