Learn More
DUBOIS CHEMICALS Surface Cleanse/930 | 1l
Catalog No. NC1720316
Encompass
A powerful, nonionic liquid cleaner designed for critical cleaning applications where a neutral pH is required.
Catalog No. NC1720316 Supplier DUBOIS CHEMICALS Supplier No. S200112
A powerful, nonionic liquid cleaner designed for critical cleaning applications where a neutral pH is required.