Learn More
Mopec Surface GSR Collection Kit
SDP
Catalog No. 22446268 Shop All Mopec Products
Change view
Click to view available options
Quantity:
10/Cs.
Catalog No. Quantity
22-446-268 10/Cs.
1 options
Catalog No. 22-446-268 Supplier Mopec Supplier No. FS483
Add to Cart
Add to Cart
Designed to collect gunshot residue from surfaces such as tabletops, vehicles, doorframes, and clothing, these materials include labeled SEM collection vials with adhesive carbon stubs. The supplied components support clear identification and traceable evidence collection for later analysis.
Construction
  • (1) Tamper-evident, pre-sealed kit envelope
  • Kit instruction sheet
  • (1) Surface gunshot residue analysis information form
  • (1) Pair of disposable plastic gloves
  • (1) Sample numbers 1-4 vials with caps containing SEM stubs covered with double-sided adhesive carbon tape
  • (1) Tamper-evident police evidence seal for resealing kit after evidence collection
Performance
  • Collects GSR from surfaces other than the suspect’s hand
Dimensions
  • Envelope: 6 L x 9 W in.
Quantity
  • 10 per case
Specifications
Product Type GSR Collection Kit
Quantity 10/Cs.