Learn More
Electron Microscopy Sciences EMS Surface Mount Tweezers Style SM 110, 120 mm
SDP
Catalog No. 5023917
Encompass_Preferred
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Straight ‘T' shaped tips with a vertical grooved inside for holding cylindrical objects up to 1mm diameter.
Catalog No. 50-239-17 Supplier Electron Microscopy Sciences Supplier No. 78250100
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Straight ‘T' shaped tips with a vertical grooved inside for holding cylindrical objects up to 1mm diameter.