Learn More
Electron Microscopy Sciences EMS Surface Mount Tweezers Style SM 112, 120 mm
SDP
Catalog No. 5023913
Encompass_Preferred
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Tips are bent and cut on a 45 degree angle (2mm edge). For perfect control (anti-capillary).
Catalog No. 50-239-13 Supplier Electron Microscopy Sciences Supplier No. 7825060
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Tips are bent and cut on a 45 degree angle (2mm edge). For perfect control (anti-capillary).