Learn More
Electron Microscopy Sciences EMS Surface Mount Tweezers Style SM 115, 120 mm
SDP
Catalog No. 5023914
Encompass_Preferred
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Very fine rounded tips with groove inside on a 30-degree angle. For positioning cylindrical objects.
Catalog No. 50-239-14 Supplier Electron Microscopy Sciences Supplier No. 7825070
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Very fine rounded tips with groove inside on a 30-degree angle. For positioning cylindrical objects.