Learn More
Electron Microscopy Sciences EMS Surface Mount Tweezers Style SM 107, 120 mm
SDP
Catalog No. 5023918
Encompass_Preferred
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Flat tip 60 degrees angle, the tips measure 1.2mm wide, for positioning all flat devices,
Catalog No. 50-239-18 Supplier Electron Microscopy Sciences Supplier No. 7825090
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. They offer comfort and smooth handling and have a satin antiglare finish with an ergonomic design. Flat tip 60 degrees angle, the tips measure 1.2mm wide, for positioning all flat devices,