Learn More
MSC SPI Surface Plate Cleaner, Granite, Pump Spray, 16 oz
Catalog No. 502866282
Encompass
This granite surface plate cleaner is formulated for the effective cleaning, degreasing, and care of granite surfaces. It is supplied in a convenient pump spray bottle for ease of application.
  • Designed for cleaning, degreasing, and care of granite surfaces
  • Features a clear thin liquid appearance
  • Water soluble for easy cleanup
  • Has a pH of 12.8
  • Combustible liquid with a flash point of 170°F (76.7°C)
Catalog No. 50-286-6282 Supplier MSC Supplier No. 00152017
This granite surface plate cleaner is formulated for the effective cleaning, degreasing, and care of granite surfaces. It is supplied in a convenient pump spray bottle for ease of application.
  • Designed for cleaning, degreasing, and care of granite surfaces
  • Features a clear thin liquid appearance
  • Water soluble for easy cleanup
  • Has a pH of 12.8
  • Combustible liquid with a flash point of 170°F (76.7°C)