Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ZEP Surface Protectant, Liquid, Protective Coating, 5 gal

Catalog No. 502867295
Encompass

This surface protectant is a milky, white liquid designed for use as a protective coating. It comes in a 5-gallon pail and is stable under normal conditions, offering a pH range of 5.5-6.5.

  • Liquid form
  • Milky white color with a slight odor
  • Packaged in a 5 gal pail
  • Stable under normal conditions
  • Soluble in water and other solvents
  • Has a pH between 5.5 and 6.5
  • Density of 0.993 g/cm3

Catalog No. 50-286-7295 Supplier MSC Supplier No. 17159195
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This surface protectant is a milky, white liquid designed for use as a protective coating. It comes in a 5-gallon pail and is stable under normal conditions, offering a pH range of 5.5-6.5.

  • Liquid form
  • Milky white color with a slight odor
  • Packaged in a 5 gal pail
  • Stable under normal conditions
  • Soluble in water and other solvents
  • Has a pH between 5.5 and 6.5
  • Density of 0.993 g/cm3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.