Learn More
BEST SANITIZERS Alpet D2 Surface Sanitizing Wipes, 160 ct. Medium Duty
Catalog No. 50828720
Encompass_Preferred
This item has a minimum order quantity of 2 cases per supplier requirements
Alpet D2 Surface Sanitizing Wipes are a ready-to-use, no-rinse, lintless wiping system for food contact surfaces. Alpet D2 Surface Sanitizing Wipes are available in a 160 count medium duty wipe. As with all Best Sanitizers wiping systems, the spring-loaded lid closes on its own to prevent the drying of wipes, helping to eliminate unnecessary waste. Kills 99.999% of Staphylococcus aureus and Escherichia coli on hard, non-porous surfaces in 60 seconds. Strong, lintless wipes for food contact surfaces Spring-loaded lid to prevent drying Ready-to-use, no-rinse required 58% IPA/quat formula ideal for water sensitive equipment NSF listed D2 classification Kosher, Pareve and Halal certified
This item is not returnable. View return policy
Catalog No. 50-828-720 Supplier BEST SANITIZERS Supplier No. SSW0002
This item has a minimum order quantity of 2 cases per supplier requirements
Alpet D2 Surface Sanitizing Wipes are a ready-to-use, no-rinse, lintless wiping system for food contact surfaces. Alpet D2 Surface Sanitizing Wipes are available in a 160 count medium duty wipe. As with all Best Sanitizers wiping systems, the spring-loaded lid closes on its own to prevent the drying of wipes, helping to eliminate unnecessary waste. Kills 99.999% of Staphylococcus aureus and Escherichia coli on hard, non-porous surfaces in 60 seconds. Strong, lintless wipes for food contact surfaces Spring-loaded lid to prevent drying Ready-to-use, no-rinse required 58% IPA/quat formula ideal for water sensitive equipment NSF listed D2 classification Kosher, Pareve and Halal certified