Learn More
BEST SANITIZERS Alpet D2 Surface Sanitizer / Disinfectant, Quart Bottles
Supplier:  BEST SANITIZERS SS10009
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Alpet D2 Surface Sanitizer is a ready-to-use, no-rinse, fast acting, food contact and non-food contact surface cleaner and sanitizer. It is highly evaporative and ideal for water sensitive equipment and dry environments. New Claims and Uses Same Reliable Formula (58% IPA/quat formula) Clean and Sanitize with one product 60 Second Kill on food contact surfaces, including rubber gloves* 10 Second Kill on non-food contact surfaces, including rubber footwear** Kills 99.999% of 9 tested pathogens as a food contact surface sanitizer Highly Evaporative and ideal for water sensitive equipment and low moisture environments NSF Listed, EPA Registered, Kosher, Pareve and Halal *Kills 99.999% of tested pathogens in 60 seconds on food contact surfaces, including rubber gloves**Kills 99.9% of tested pathogens in 10 seconds on non-food contact surfaces, including rubber footwear
Catalog No. 50-828-719