Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Surface Tension of Water Kit
GREENER_CHOICESCI_ED

Catalog No. S37748 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Demonstrations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S37748 5 Demonstrations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37748 Supplier Innovating Science™ Supplier No. IS7012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

1 x 15 g Sulfur NA1350, 1 x 5mL Green Dish Detergent

Kit to explain principle of high surface tension of water to students.

  • Water acts like a membrane stretched across the surface preventing the sulfur particles from sinking.
  • Wetting agent lowers the surface tension and allows the particles to fall through the surface to the bottom of the beaker.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Explains principle of high surface tension of water to students
Includes 1 x 15 g Sulfur NA1350, 1 x 5 mL Green Dish Detergent
Quantity 1
Dimensions (L x W x H) 6.625 x 3.25 x 2.25 in.
Class Size 5 Demonstrations
Product Type Surface Tension of Water Kit

DOT Info: Non-Regulated.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.