Learn More
Ptc Instruments SURFACE THERM -10C TO 150C
Catalog No. NC9023621
Encompass
The 310C is 2 in. in diameter and has an accuarcy is +/- 2% of Full Scale
Catalog No. NC9023621 Supplier Ptc Instruments Supplier No. 310C
The 310C is 2 in. in diameter and has an accuarcy is +/- 2% of Full Scale