Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Surfact-Amps™ Detergent Sampler

Catalog No. PI28340 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28340 10 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI28340 Supplier Thermo Scientific™ Supplier No. 28340
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convenient collection of various Surfact-Amps Detergent Solutions and other selected useful detergents for protein research methods.

Thermo Scientific™ Surfact-Amps Detergent Sampler is a convenient collection of various Thermo Scientific™ Surfact-Amps Detergent Solutions and selected other useful detergents for protein research methods.

The sampler kit is an economical way to test which detergent is most effective for a particular protein storage or experimental system. Thermo Scientific Surfact-Amps Purified Detergent Solutions are convenient 10% (w/v) solutions of commonly used detergents that provide unsurpassed purity, quality and stability for proteomics and molecular biology methods. Unlike neat detergent formulations, Surfact-Amps 10% Solutions are not so viscous that you can't aliquot them accurately. The detergent solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products. Don't begin your experiment with detergents of unknown age and purity! Just open an ampule and dilute the contents into your buffer at the desired concentration. Three of the ten detergents in the sampler kit are supplied as solids (100mg each) rather than 10% solutions.

Highlights:

Surfact-Amps Detergent Solutions are

  • Precise 10% detergent solutions in ultrapure water ampuled under nitrogen

  • Easy to accurately dispense and dilute for use
  • Exceptionally pure: less than 1.0 μeq/mL peroxides and carbonyls
  • Highly stable: packed in glass ampules under inert nitrogen gas

Includes:

One ampule each of seven different detergent solutions and 100mg each of three additional detergent solids. Brij™, Triton™ and Tween™ are trademarks of their respective owners.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Detergent
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Liquid
Product Line Surfact-Amps
Reagent Type Detergent Solution
Quantity 10 Vials

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.