Learn More
Carbosynth LLC. Surfactant 10G for use as a detergent, wetting agent, emulsifier, foaming agent, and dispersant.
SDP
Catalog No. 501253527
Encompass_Preferred
Surfactants are compounds that lower the surface tension of a liquid, the interfacial tension between two liquids, or that between a liquid and a solid. Surfactants may act as detergents, wetting agents, emulsifiers, foaming agents, and dispersants.
Catalog No. 50-125-3527 Supplier Carbosynth LLC. Supplier No. 95R10310G
Surfactants are compounds that lower the surface tension of a liquid, the interfacial tension between two liquids, or that between a liquid and a solid. Surfactants may act as detergents, wetting agents, emulsifiers, foaming agents, and dispersants.