Learn More
Biomatik Corporation Surfactant Protein C (Sp-C) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30549
Encompass_Preferred
Surfactant Protein C (Sp-C) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Surfactant Protein C (SP-C). The Surfactant Protein C (Sp-C) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Phe24~Ile191. Applications: WB. Target Synonyms: Surfactant Protein C (SP-C). Accession Number: P11686. Gene ID: 6440. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9205