Learn More
Medchemexpress LLC Surfactin | 252023-70-4 | 98.96% | 50 MG
SDP
Catalog No. 5000339448
Encompass_Preferred
Surfactin is a potent cyclic lipopeptide biosurfactant consisting of four isomers (Surfactin A, B, C, and D). It mediates the flux of mono- and divalent cations, such as calcium, across lipid bilayer membranes. Surfactin acts as an antimicrobial adjuvant with antibacterial, antifungal, antimycoplasma, and hemolytic effects, and has antiviral activity against various enveloped viruses.
  • Potent cyclic lipopeptide biosurfactant
  • Consists of four isomers
  • Mediates flux of mono- and divalent cations across lipid bilayer membranes
  • Acts as an antimicrobial adjuvant
  • Exhibits antibacterial, antifungal, and antimycoplasma effects
  • Has antiviral activity against various enveloped viruses
Catalog No. 50-003-39448 Supplier Medchemexpress LLC Supplier No. HY12955550MG
Surfactin is a potent cyclic lipopeptide biosurfactant consisting of four isomers (Surfactin A, B, C, and D). It mediates the flux of mono- and divalent cations, such as calcium, across lipid bilayer membranes. Surfactin acts as an antimicrobial adjuvant with antibacterial, antifungal, antimycoplasma, and hemolytic effects, and has antiviral activity against various enveloped viruses.
  • Potent cyclic lipopeptide biosurfactant
  • Consists of four isomers
  • Mediates flux of mono- and divalent cations across lipid bilayer membranes
  • Acts as an antimicrobial adjuvant
  • Exhibits antibacterial, antifungal, and antimycoplasma effects
  • Has antiviral activity against various enveloped viruses