Learn More
Cayman Chemical Surfactn 100ug
SDP
Catalog No. 502988013
Encompass_Preferred
A cyclic lipopeptide biosurfactant; composed of a variable heptapeptide and a β-hydroxy fatty acid that ranges in length from 12- to 15 carbons; contains the heptapeptide L-Glu-L-Leu-D-Leu-L-Val-L-Asp-D-Leu-L-Leu and is linked to a 15-carbon β-hydroxy fatty acid with the iso branching configuration
Catalog No. 50-298-8013 Supplier Cayman Chemical Supplier No. 31620100 UG
A cyclic lipopeptide biosurfactant; composed of a variable heptapeptide and a β-hydroxy fatty acid that ranges in length from 12- to 15 carbons; contains the heptapeptide L-Glu-L-Leu-D-Leu-L-Val-L-Asp-D-Leu-L-Leu and is linked to a 15-carbon β-hydroxy fatty acid with the iso branching configuration