Learn More
Cayman Chemical Surfactnt ProteIn B RabbIt M
SDP
Catalog No. 502949375
Encompass_Preferred
An internal standard for the quantification of tasimelteon by GC- or LC-MS
Catalog No. 50-294-9375 Supplier Cayman Chemical Supplier No. 32295100 UL
An internal standard for the quantification of tasimelteon by GC- or LC-MS