Learn More
Cayman Chemical Surfen (hydrochloride), Purity: ≥98%, CAS: 5424-37-3, 10 mg
SDP
Catalog No. 502936923
Encompass_Preferred
An antagonist of cell surface glycosaminoglycans; binds to heparin, dermatan sulfate, heparan sulfate, and chondroitin sulfate and prevents heparin-induced inhibition of Factor Xa in cell-free assays; inhibits FGF2 binding to CHO cells (IC50 = 5 µM) and FGF2-induced tube formation by primary mouse endothelial cells; inhibits neural differentiation and increases pluripotency in mESCs at 5 µM
Catalog No. 50-293-6923 Supplier Cayman Chemical Supplier No. 2667010 MG
An antagonist of cell surface glycosaminoglycans; binds to heparin, dermatan sulfate, heparan sulfate, and chondroitin sulfate and prevents heparin-induced inhibition of Factor Xa in cell-free assays; inhibits FGF2 binding to CHO cells (IC50 = 5 µM) and FGF2-induced tube formation by primary mouse endothelial cells; inhibits neural differentiation and increases pluripotency in mESCs at 5 µM