Learn More
Med Vet International 3M Surgical Clipper Kit Pivoting Head 160 Minute Run Time, 9667L, 1 Kit
Catalog No. 502831727
Encompass_Preferred
This 3M™ hair clipper shaves away hair prior to surgery A pivoting head navigates around creases, folds and contours of skin for more efficient use The blade attachment ensures maximum skin contact for easier, more productive use This 3M™ surgical clipper starter kit helps prevent nicks and cuts with its away-from-skin cutting position Kit includes drop-in stand for convenient storage The handle rinses clean easily for quick maintenance Each four-hour charge of this clipper's lithium-ion battery provides 160 minutes of run time
Catalog No. 50-283-1727 Supplier Med Vet International Supplier No. 1061817KT
This 3M™ hair clipper shaves away hair prior to surgery A pivoting head navigates around creases, folds and contours of skin for more efficient use The blade attachment ensures maximum skin contact for easier, more productive use This 3M™ surgical clipper starter kit helps prevent nicks and cuts with its away-from-skin cutting position Kit includes drop-in stand for convenient storage The handle rinses clean easily for quick maintenance Each four-hour charge of this clipper's lithium-ion battery provides 160 minutes of run time