Learn More
Ace Glass, Inc. SURGE FLASK SURGE FLASK
SDP
Catalog No. 50975463
Encompass_Preferred
3L Surge Flask with a 45/50 outer joint and a #11 thread for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-463 Supplier Ace Glass, Inc. Supplier No. 781826
3L Surge Flask with a 45/50 outer joint and a #11 thread for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.