Learn More
AIMS™ Surgery Board System for Small Animals
SCI_ED
Catalog No. 14370213
Change view
Click to view available options
Includes:
Magnetic Base System
No Magnetic Base System
Dimensions (L x W):
30.48 x 20.42 cm
30.48 x 24.82 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Dimensions (L x W)
14-370-213 No Magnetic Base System 30.48 x 20.42 cm
14-370-210 Magnetic Base System 30.48 x 24.82 cm
14-370-211 No Magnetic Base System 30.48 x 24.82 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-370-213 Supplier AIMS™ Supplier No. GSS1
Add to Cart
Add to Cart
For small animal surgery, procedure, and dissection platforms.
  • Construction: High Density Polyethylene (HDPE) with excellent chemical resistance
  • Corner magnets for holding metal accessories
  • Recessed Steel Bars for use with magnetic accessories
  • Flexible LED lights - provides direct illu
Specifications
Animal Mouse, Rat
For Use With (Application) Dissection of Small Animals
Type Surgical Board
Includes No Magnetic Base System
Magnification 2.3X (main lens); 4.0X (inset lens)
Material HDPE, Stainless Steel (Warming Plate)
Dimensions (L x W) 30.48 x 20.42 cm
Electrical Requirements 100/240 V 50/60 Hz