Learn More
Sklar Instruments Surgi-OR Babcock Forceps, Sklar 8 in., Fenestrated, Straight, Stainless Steel. EA.
SDP
Catalog No. 501184256 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Babcock Forceps, Sklar. 8 in., Fenestrated, Straight, Stainless Steel. Surgi-OR Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue. They are frequently used with intestinal and laparotomy procedures. Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface. The jaws are circumferential, and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight and the overall length is 8 inches.
Catalog No. 50-118-4256 Supplier Sklar Instruments Supplier No. 95956
Surgi-OR Babcock Forceps, Sklar. 8 in., Fenestrated, Straight, Stainless Steel. Surgi-OR Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue. They are frequently used with intestinal and laparotomy procedures. Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface. The jaws are circumferential, and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight and the overall length is 8 inches.