Learn More
Sklar Instruments Forceps, Surgi-OR Adson Bayonet Dressing Forceps, Sklar 4-3/4 in., Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126562 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Surgi-OR Adson Bayonet Dressing Forceps, Sklar, 4-3/4 in., Straight, Serrated, Stainless Steel, EA. Surgi-OR Adson Bayonet Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Surgi-OR mid-grade instruments provide cost-effective, reliable instrumentation for the operating room and surgical centers. This product is straight with serrated tips and a length of 4-3/4 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6562 Supplier Sklar Instruments Supplier No. 95771
Forceps, Surgi-OR Adson Bayonet Dressing Forceps, Sklar, 4-3/4 in., Straight, Serrated, Stainless Steel, EA. Surgi-OR Adson Bayonet Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Surgi-OR mid-grade instruments provide cost-effective, reliable instrumentation for the operating room and surgical centers. This product is straight with serrated tips and a length of 4-3/4 inches. Lifetime warranty against all manufacturer defects.