Learn More
Sklar Instruments Surgi-OR Joseph Hook, Sklar 6-1/4 in., 2 Sharp Prongs, 2mm, Straight, Stainless Steel. EA.
SDP
Catalog No. 501184210 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Joseph Hook, Sklar. 6-1/4 in., 2 Sharp Prongs, 2mm, Straight, Stainless Steel. The Surgi-OR Joseph Skin Hook is a handheld retractor commonly used for holding back the edges of skin during intranasal and pharyngeal procedures. It is available with a single or dual prong, sharp hook. The handle is often round and knurled and it tapers to the hook end. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with a sharp, double prong, hook tip and a length of 6-1/4 inches. The space between the two prongs is 2mm.
Catalog No. 50-118-4210 Supplier Sklar Instruments Supplier No. 95392
Surgi-OR Joseph Hook, Sklar. 6-1/4 in., 2 Sharp Prongs, 2mm, Straight, Stainless Steel. The Surgi-OR Joseph Skin Hook is a handheld retractor commonly used for holding back the edges of skin during intranasal and pharyngeal procedures. It is available with a single or dual prong, sharp hook. The handle is often round and knurled and it tapers to the hook end. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with a sharp, double prong, hook tip and a length of 6-1/4 inches. The space between the two prongs is 2mm.