Learn More
Sklar Instruments Forceps, Surgi-OR Plain Splinter Forceps, Sklar 3-1/2 in., Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126565 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Surgi-OR Plain Splinter Forceps, Sklar, 3-1/2 in., Straight, Serrated, Stainless Steel, EA. Surgi-OR Plain Splinter Forceps are multi-purpose thumb forceps typically used for grasping and removing small foreign objects with their fine, pointed tips. They may also be used for holding bandages, sutures, or other items. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with serrated jaws and pointed tips. It has a length of 3-1/2 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6565 Supplier Sklar Instruments Supplier No. 95777
Forceps, Surgi-OR Plain Splinter Forceps, Sklar, 3-1/2 in., Straight, Serrated, Stainless Steel, EA. Surgi-OR Plain Splinter Forceps are multi-purpose thumb forceps typically used for grasping and removing small foreign objects with their fine, pointed tips. They may also be used for holding bandages, sutures, or other items. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with serrated jaws and pointed tips. It has a length of 3-1/2 inches. Lifetime warranty against all manufacturer defects.